Product Details

SNP ID
rs16859317
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184816851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGACCATCTCCTATTCTGTGC[C/T]CACTGTGAAGCAAGAAGTTATTGAG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
VPS8 PubMed Links
Additional Information
For this assay, SNP(s) [rs74921343] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
VPS8
Gene Name
VPS8, CORVET complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009921.2 Intron NP_001009921.1
NM_015303.3 Intron NP_056118.2
XM_005247251.3 Intron XP_005247308.1
XM_005247253.4 Intron XP_005247310.1
XM_006713556.3 Intron XP_006713619.1
XM_011512599.2 Intron XP_011510901.1
XM_011512600.2 Intron XP_011510902.1
XM_011512601.2 Intron XP_011510903.1
XM_017006040.1 Intron XP_016861529.1
XM_017006041.1 Intron XP_016861530.1

View Full Product Details