Product Details

SNP ID
rs17104901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:76827366 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCCACCATCCTAGAATCTGGGTG[A/G]TCAAAACTTGGGACCCCTGCCACAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC74A PubMed Links

Gene Details

Gene
LRRC74A
Gene Name
leucine rich repeat containing 74A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194287.2 503 Intron NP_919263.2
XM_011536479.1 503 Intron XP_011534781.1
XM_011536483.1 503 Intron XP_011534785.1
XM_011536487.1 503 Intron XP_011534789.1
XM_017021032.1 503 Intron XP_016876521.1
XM_017021033.1 503 Intron XP_016876522.1
XM_017021034.1 503 Intron XP_016876523.1
XM_017021035.1 503 Intron XP_016876524.1
XM_017021036.1 503 Intron XP_016876525.1
XM_017021037.1 503 UTR 5 XP_016876526.1
XM_017021038.1 503 Intron XP_016876527.1
XM_017021039.1 503 Intron XP_016876528.1
XM_017021040.1 503 Intron XP_016876529.1
XM_017021041.1 503 Intron XP_016876530.1

View Full Product Details