Product Details

SNP ID
rs17695669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:18664928 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTGAGTTGCTTCAAGAAGGAGCC[A/G]TCTGGTACTGTCCTTCCATTCCTTT
Phenotype
MIM: 608909
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL5B PubMed Links

Gene Details

Gene
ARL5B
Gene Name
ADP ribosylation factor like GTPase 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178815.3 Intron NP_848930.1
XM_005252400.1 Intron XP_005252457.1
XM_005252401.4 Intron XP_005252458.1
Gene
NSUN6
Gene Name
NOP2/Sun RNA methyltransferase family member 6
There are no transcripts associated with this gene.

View Full Product Details