Product Details

SNP ID
rs34107519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1762803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTAGGCCGGCAGGTGTATAGGGC[A/G]TGGGGGCCCAGCTGCCAGTGCTGGA
Phenotype
MIM: 172860 MIM: 613168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINF1 PubMed Links

Gene Details

Gene
SERPINF1
Gene Name
serpin family F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017024754.1 Intron XP_016880243.1
Gene
SERPINF2
Gene Name
serpin family F member 2
There are no transcripts associated with this gene.

View Full Product Details