Product Details

SNP ID
rs17313882
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:84065208 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGACAGCATCAATAGCCTTGCAAT[C/G]TGTTTATTTCACCTACATTTATGCA
Phenotype
MIM: 300303
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RPS6KA6 PubMed Links

Gene Details

Gene
RPS6KA6
Gene Name
ribosomal protein S6 kinase A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014496.4 Intron NP_055311.1
XM_011530917.2 Intron XP_011529219.1
XM_011530918.2 Intron XP_011529220.1
XM_011530919.2 Intron XP_011529221.1
XM_011530920.2 Intron XP_011529222.1
XM_017029423.1 Intron XP_016884912.1
XM_017029424.1 Intron XP_016884913.1
XM_017029425.1 Intron XP_016884914.1
XM_017029426.1 Intron XP_016884915.1

View Full Product Details