Product Details

SNP ID
rs16980188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:85277795 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTAAGTATTTTGGAAAACTCACA[C/T]CTGTTCTCATTTTTAAACCTTAATA
Phenotype
MIM: 300603 MIM: 314990
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POF1B PubMed Links

Gene Details

Gene
POF1B
Gene Name
premature ovarian failure, 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307940.1 3455 Intron NP_001294869.1
NM_024921.3 3455 UTR 3 NP_079197.3
XM_005262203.3 3455 UTR 3 XP_005262260.1
Gene
ZNF711
Gene Name
zinc finger protein 711
There are no transcripts associated with this gene.

View Full Product Details