Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001010923.2 | 11596 | Intron | NP_001010923.1 | ||
NM_001164685.1 | 11596 | Intron | NP_001158157.1 | ||
NM_001164687.1 | 11596 | Intron | NP_001158159.1 | ||
NM_001318531.1 | 11596 | Intron | NP_001305460.1 | ||
XM_011535813.2 | 11596 | UTR 3 | XP_011534115.1 | ||
XM_011535814.2 | 11596 | Intron | XP_011534116.1 | ||
XM_011535816.1 | 11596 | Intron | XP_011534118.1 | ||
XM_017010848.1 | 11596 | Intron | XP_016866337.1 | ||
XM_017010849.1 | 11596 | Intron | XP_016866338.1 |