Product Details

SNP ID
rs16988594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:31879397 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGATTGCATTCTGCAGTTGCAGGC[A/G]GATTCTTTTGAACTTCCTGCCCTGC
Phenotype
MIM: 606532
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HUNK PubMed Links

Gene Details

Gene
HUNK
Gene Name
hormonally up-regulated Neu-associated kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014586.1 Intron NP_055401.1
XM_011529537.2 Intron XP_011527839.1

View Full Product Details