Product Details

SNP ID
rs17743655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29306431 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTCCCTAAAGCCTCATAGCAGA[C/T]GCTACCTTGTCCCTCTCTCCTCTCT
Phenotype
MIM: 602751
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BACH1 PubMed Links

Gene Details

Gene
BACH1
Gene Name
BTB domain and CNC homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001186.3 Intron NP_001177.1
NM_206866.2 Intron NP_996749.1

View Full Product Details