Product Details
- SNP ID
-
rs17860250
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:33233339 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAATTGGGACCACTGAAGTCCCA[C/G]ATGTAGAGAAGTCAACAAATGGGAC
- Phenotype
-
MIM: 602376
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IFNAR2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17860130] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IFNAR2
- Gene Name
- interferon alpha and beta receptor subunit 2
View Full Product Details