Product Details

SNP ID
rs17860249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33231536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTAAAAGTCATTTCAAAAAGTA[A/G]TATGAAAAGGATCATAACCCTAGAG
Phenotype
MIM: 602376
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFNAR2 PubMed Links

Gene Details

Gene
IFNAR2
Gene Name
interferon alpha and beta receptor subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000874.4 Intron NP_000865.2
NM_001289125.1 Intron NP_001276054.1
NM_001289126.1 Intron NP_001276055.1
NM_001289128.1 Intron NP_001276057.1
NM_207584.2 Intron NP_997467.1
NM_207585.2 Intron NP_997468.1

View Full Product Details