Product Details

SNP ID
rs11540070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:10038320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAGCATGGCTTTAGCAGATAAG[A/G]GACTTGAGAACTTACAGATCTACAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKEF1 PubMed Links

Gene Details

Gene
ANKEF1
Gene Name
ankyrin repeat and EF-hand domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303472.1 462 UTR 5 NP_001290401.1
NM_022096.5 462 Missense Mutation AGA,GGA R7G NP_071379.3
NM_198798.2 462 Missense Mutation AGA,GGA R7G NP_942093.1
XM_005260792.2 462 Intron XP_005260849.1
Gene
SNAP25-AS1
Gene Name
SNAP25 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details