Product Details

SNP ID
rs16851751
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:166196876 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATTGAGTTTTATACGTAGATTAG[A/G]ACAAATGTCTCAAAGTAATAACGGA
Phenotype
MIM: 603415
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LOC101929680 PubMed Links

Gene Details

Gene
LOC101929680
Gene Name
uncharacterized LOC101929680
There are no transcripts associated with this gene.

Gene
SCN9A
Gene Name
sodium voltage-gated channel alpha subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002977.3 8071 UTR 3 NP_002968.1
XM_005246757.2 8071 UTR 3 XP_005246814.1
XM_011511616.2 8071 Intron XP_011509918.1
XM_011511617.2 8071 Intron XP_011509919.1
XM_011511618.2 8071 Intron XP_011509920.1
XM_011511619.2 8071 Intron XP_011509921.1
XM_017004668.1 8071 Intron XP_016860157.1
XM_017004669.1 8071 Intron XP_016860158.1

View Full Product Details