Product Details

SNP ID
hCV42486049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:149900267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGCAGTGAGACTCACTTCCGC[G/T]TGGCTGTGGTGAGCTCTCGTTTCGA
Phenotype
MIM: 613181 MIM: 185860
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BOLA1 PubMed Links

Gene Details

Gene
BOLA1
Gene Name
bolA family member 1
There are no transcripts associated with this gene.

Gene
SV2A
Gene Name
synaptic vesicle glycoprotein 2A
There are no transcripts associated with this gene.

View Full Product Details