Product Details

SNP ID
rs4500305
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:40784354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACCGCCGCCTGCAGAACTGGGT[C/G]TACAACGTGCTGGAGCGGCCCCGCG
Phenotype
MIM: 603537
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCNQ4 PubMed Links

Gene Details

Gene
KCNQ4
Gene Name
potassium voltage-gated channel subfamily Q member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004700.3 343 Silent Mutation NP_004691.2
NM_172163.2 343 Silent Mutation NP_751895.1
XM_017002792.1 343 Intron XP_016858281.1

View Full Product Details