Product Details

SNP ID
rs11538424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219178604 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAGAAGCCGCTGCACAGCCTGGT[C/T]ACGGCGGCCGCGCTCAACGGCCTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNPPD1 PubMed Links

Gene Details

Gene
CNPPD1
Gene Name
cyclin Pas1/PHO80 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321389.1 415 Intron NP_001308318.1
NM_001321390.1 415 Intron NP_001308319.1
NM_001321391.1 415 Intron NP_001308320.1
NM_015680.5 415 Intron NP_056495.3
Gene
FAM134A
Gene Name
family with sequence similarity 134 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321109.1 415 UTR 5 NP_001308038.1
NM_001321110.1 415 Intron NP_001308039.1
NM_024293.5 415 Silent Mutation GTC,GTT V84V NP_077269.3
XM_005246848.2 415 Silent Mutation GTC,GTT V84V XP_005246905.1
Gene
SLC23A3
Gene Name
solute carrier family 23 member 3
There are no transcripts associated with this gene.

View Full Product Details