Product Details

SNP ID
rs11556310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219539435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGTAGCATGGCCAGCTGGGTT[C/T]GGGAGCCGAGGCCCTCAAGCACGCT
Phenotype
MIM: 606715 MIM: 610405 MIM: 610991
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASIC4 PubMed Links

Gene Details

Gene
ASIC4
Gene Name
acid sensing ion channel subunit family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182847.2 2034 Intron NP_878267.2
XM_005246671.2 2034 Intron XP_005246728.2
XM_017004439.1 2034 Intron XP_016859928.1
Gene
CHPF
Gene Name
chondroitin polymerizing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195731.1 2034 Missense Mutation CAA,CGA Q597R NP_001182660.1
NM_024536.5 2034 Missense Mutation CAA,CGA Q759R NP_078812.2
XM_011511838.2 2034 Missense Mutation CAA,CGA Q468R XP_011510140.1
Gene
MIR3132
Gene Name
microRNA 3132
There are no transcripts associated with this gene.

Gene
OBSL1
Gene Name
obscurin like 1
There are no transcripts associated with this gene.

Gene
TMEM198
Gene Name
transmembrane protein 198
There are no transcripts associated with this gene.

View Full Product Details