Product Details

SNP ID
rs6725330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:226802141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCACGTACCTCACAATTATCTTC[A/G]TAGGAAGGATTTTGATCATAAAAAG
Phenotype
MIM: 147545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRS1 PubMed Links

Gene Details

Gene
IRS1
Gene Name
insulin receptor substrate 1
There are no transcripts associated with this gene.

Gene
RHBDD1
Gene Name
rhomboid domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167608.1 Intron NP_001161080.1
NM_032276.3 Intron NP_115652.2
XM_017005083.1 Intron XP_016860572.1
XM_017005084.1 Intron XP_016860573.1
XM_017005085.1 Intron XP_016860574.1
XM_017005086.1 Intron XP_016860575.1
XM_017005087.1 Intron XP_016860576.1
XM_017005088.1 Intron XP_016860577.1
XM_017005089.1 Intron XP_016860578.1
XM_017005090.1 Intron XP_016860579.1
XM_017005091.1 Intron XP_016860580.1
XM_017005092.1 Intron XP_016860581.1
XM_017005093.1 Intron XP_016860582.1
XM_017005094.1 Intron XP_016860583.1
XM_017005095.1 Intron XP_016860584.1

View Full Product Details