Product Details

SNP ID
rs11549990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179570192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTACATAGATACTAATGCTCTGCG[A/T]GTTCCGAGGGAGAATATGGAGGCCA
Phenotype
MIM: 604958
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ACTL6A PubMed Links

Gene Details

Gene
ACTL6A
Gene Name
actin like 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004301.4 489 Silent Mutation CGA,CGT R76R NP_004292.1
NM_177989.3 489 Silent Mutation CGA,CGT R34R NP_817126.1
NM_178042.3 489 Silent Mutation CGA,CGT R34R NP_829888.1

View Full Product Details