Product Details

SNP ID
rs11567693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:35857364 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAATGCTCATGTAGATGCTATGAC[G/T]GTACTTGTAGGAAAATGTCCAAAGT
Phenotype
MIM: 146661
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IL7R PubMed Links

Gene Details

Gene
IL7R
Gene Name
interleukin 7 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002185.3 Intron NP_002176.2
XM_005248299.3 Intron XP_005248356.1

View Full Product Details