Product Details

SNP ID
rs6884331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:36252850 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACTTACACCTAACAGTACACTTA[C/T]TGGTAGCTAATAAACTTTGGTTTTT
Phenotype
MIM: 616391
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RANBP3L PubMed Links

Gene Details

Gene
RANBP3L
Gene Name
RAN binding protein 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161429.2 Intron NP_001154901.1
NM_001323273.1 Intron NP_001310202.1
NM_001323274.1 Intron NP_001310203.1
NM_001323275.1 Intron NP_001310204.1
NM_001323276.1 Intron NP_001310205.1
NM_001323277.1 Intron NP_001310206.1
NM_001323278.1 Intron NP_001310207.1
NM_001323279.1 Intron NP_001310208.1
NM_001323280.1 Intron NP_001310209.1
NM_145000.4 Intron NP_659437.3
XM_006714452.3 Intron XP_006714515.1
XM_006714453.3 Intron XP_006714516.1
XM_006714455.3 Intron XP_006714518.1

View Full Product Details