Product Details
- SNP ID
-
rs12516635
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:75853804 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTACGTTCTTTTCCTAAATTTTG[A/T]AAAGACTGAGTCTTTGAGACCATTT
- Phenotype
-
MIM: 610291
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SV2C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78995753] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SV2C
- Gene Name
- synaptic vesicle glycoprotein 2C
View Full Product Details