Product Details
- SNP ID
-
rs1574197
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:80654908 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCGGCTGCAGCGGCCGCAGCGGCC[C/G]CAGCGCCCCCAGCGCCCCCAGCTCC
- Phenotype
-
MIM: 126060
MIM: 600887
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DHFR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2001675] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DHFR
- Gene Name
- dihydrofolate reductase
- Gene
- MSH3
- Gene Name
- mutS homolog 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002439.4 |
261 |
Missense Mutation |
CCA,GCA |
P61A |
NP_002430.3 |
- Gene
- MTRNR2L2
- Gene Name
- MT-RNR2-like 2
There are no transcripts associated with this gene.
View Full Product Details