Product Details

SNP ID
hCV43019288
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73686703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGAGCTTCTTTATCTGCCAGAG[A/C]ATAAGCCCTGTTACCTGCTGGATAT
Phenotype
MIM: 615733
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
DNAJC30 PubMed Links

Gene Details

Gene
DNAJC30
Gene Name
DnaJ heat shock protein family (Hsp40) member C30
There are no transcripts associated with this gene.

Gene
WBSCR22
Gene Name
Williams-Beuren syndrome chromosome region 22
There are no transcripts associated with this gene.

View Full Product Details