Product Details

SNP ID
rs10951788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:45008320 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCGAAGTCCTGACCAATGTGTGCC[A/T]CTGTGTGCCACCGTGCCTGGCCAAG
Phenotype
MIM: 607929
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCM2 PubMed Links

Gene Details

Gene
CCM2
Gene Name
CCM2 scaffolding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029835.2 Intron NP_001025006.1
NM_001167934.1 Intron NP_001161406.1
NM_001167935.1 Intron NP_001161407.1
NM_031443.3 Intron NP_113631.1
XM_006715785.3 Intron XP_006715848.1
XM_006715786.3 Intron XP_006715849.1
XM_011515561.2 Intron XP_011513863.1
XM_011515562.1 Intron XP_011513864.1
XM_011515563.2 Intron XP_011513865.1
XM_011515564.2 Intron XP_011513866.1
XM_017012671.1 Intron XP_016868160.1
XM_017012672.1 Intron XP_016868161.1
XM_017012673.1 Intron XP_016868162.1

View Full Product Details