Product Details

SNP ID
rs6951185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:102396430 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTGCCATGGCTAGCCCAGCCGCC[G/T]CCTCGGTGCGACCACCGAGGCCCAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100630923 PubMed Links

Gene Details

Gene
LOC100630923
Gene Name
LOC100289561-PRKRIP1 readthrough
There are no transcripts associated with this gene.

Gene
MIR548O
Gene Name
microRNA 548o
There are no transcripts associated with this gene.

Gene
PRKRIP1
Gene Name
PRKR interacting protein 1 (IL11 inducible)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024653.3 74 Missense Mutation GCC,TCC A7S NP_078929.1

View Full Product Details