Product Details

SNP ID
rs6981159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:53977440 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAACGAGGTGGCCACAGGCAAAAGT[A/G]GGAATGGAATTTCCTACTCATAAAC
Phenotype
MIM: 601425
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCEA1 PubMed Links

Gene Details

Gene
TCEA1
Gene Name
transcription elongation factor A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006756.3 Intron NP_006747.1
NM_201437.2 Intron NP_958845.1
XM_006716467.1 Intron XP_006716530.1
XM_011517578.1 Intron XP_011515880.1

View Full Product Details