Product Details
- SNP ID
-
rs7341737
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:5454014 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTTCTTTTCTTATTTTCTGTAATT[C/T]CATTATATTACTTTAATAATAAAGA
- Phenotype
-
MIM: 605402
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD274
PubMed Links
Gene Details
- Gene
- CD274
- Gene Name
- CD274 molecule
View Full Product Details