Product Details

SNP ID
rs7123156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3627116 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATAGCAGAAAAAGCCCTTGGTTGG[C/G]AGTTCCAGTCCCAGTTCTGTCAGTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRPC2 PubMed Links

Gene Details

Gene
TRPC2
Gene Name
transient receptor potential cation channel subfamily C member 2, pseudogene
There are no transcripts associated with this gene.

View Full Product Details