Product Details

SNP ID
rs5743216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:43319380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTCTTGTCTTTTCAGATGAAAC[A/G]GCGAATTGCCAACATTTCATGCAGA
Phenotype
MIM: 609774
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
API5 PubMed Links

Gene Details

Gene
API5
Gene Name
apoptosis inhibitor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142930.1 Intron NP_001136402.1
NM_001142931.1 Intron NP_001136403.1
NM_001243747.1 Intron NP_001230676.1
NM_006595.3 Intron NP_006586.1
XM_006718359.3 Intron XP_006718422.1
XM_017018464.1 Intron XP_016873953.1
XM_017018465.1 Intron XP_016873954.1

View Full Product Details