Product Details
- SNP ID
-
rs745324520
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:73875957 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAAGGAGCCCAGATACACATTCT[C/T]TGATTTCTAGCATAGGCAGGTAGGT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TBC1D21
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77891899] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TBC1D21
- Gene Name
- TBC1 domain family member 21
View Full Product Details