Product Details
- SNP ID
-
rs7186210
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:24256090 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGGGGCAACAGGCGGGGAGCTGTC[C/T]CTTCAGCACCACGGACCTTGGCGCC
- Phenotype
-
MIM: 606403
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CACNG3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138438483] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CACNG3
- Gene Name
- calcium voltage-gated channel auxiliary subunit gamma 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006539.3 |
538 |
UTR 5 |
|
|
NP_006530.1 |
View Full Product Details