Product Details

SNP ID
rs8049458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46587537 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAACTCACTTCAAACAGAATATA[C/G]GCAAGTTGAAATTAAAACAATAGAA
Phenotype
MIM: 611027
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SHCBP1 PubMed Links

Gene Details

Gene
SHCBP1
Gene Name
SHC binding and spindle associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324318.1 Intron NP_001311247.1
NM_001324319.1 Intron NP_001311248.1
NM_024745.4 Intron NP_079021.3
XM_011523335.2 Intron XP_011521637.1

View Full Product Details