Product Details

SNP ID
rs7201175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88426827 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGCCCAGGCCCTGAGAGGTTCAGG[C/T]TGAGCCTCTGCTGCCTCAGTCCCCT
Phenotype
MIM: 612078
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF469 PubMed Links

Gene Details

Gene
ZNF469
Gene Name
zinc finger protein 469
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127464.2 507 Intron NP_001120936.2
XM_017023784.1 507 UTR 5 XP_016879273.1
XM_017023785.1 507 Intron XP_016879274.1

View Full Product Details