Product Details

SNP ID
rs11546986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81869573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCAGCTCCAGGGGGCCCGGGGC[A/C]GAGCTGCACACCAGGGTCAGGCCAG
Phenotype
MIM: 601925 MIM: 176790
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARHGDIA PubMed Links

Gene Details

Gene
ARHGDIA
Gene Name
Rho GDP dissociation inhibitor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185077.2 604 Silent Mutation NP_001172006.1
NM_001185078.2 604 Silent Mutation NP_001172007.1
NM_001301240.1 604 Silent Mutation NP_001288169.1
NM_001301241.1 604 Silent Mutation NP_001288170.1
NM_001301242.1 604 Silent Mutation NP_001288171.1
NM_001301243.1 604 Silent Mutation NP_001288172.1
NM_004309.5 604 Silent Mutation NP_004300.1
XM_011523574.1 604 Silent Mutation XP_011521876.1
Gene
P4HB
Gene Name
prolyl 4-hydroxylase subunit beta
There are no transcripts associated with this gene.

View Full Product Details