Product Details
- SNP ID
-
rs8077896
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:9255099 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGACCTTTCATAGTACAGTTTTAG[C/T]GCATGGTGGGTGGGTCTAGTGGAAC
- Phenotype
-
MIM: 604203
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
STX8
PubMed Links
Gene Details
- Gene
- STX8
- Gene Name
- syntaxin 8
View Full Product Details