Product Details

SNP ID
hCV43719504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41168081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGTGGTCCTGCAGCAGGTGGTC[C/T]GGCAGCAGCTGGGGCGGCAGCAGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP4-2 PubMed Links

Gene Details

Gene
KRTAP4-2
Gene Name
keratin associated protein 4-2
There are no transcripts associated with this gene.

Gene
KRTAP4-3
Gene Name
keratin associated protein 4-3
There are no transcripts associated with this gene.

Gene
KRTAP4-4
Gene Name
keratin associated protein 4-4
There are no transcripts associated with this gene.

View Full Product Details