Product Details
- SNP ID
-
rs8112421
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:753715 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACAAAAAAAAATGCTGCTGGTTGG[A/T]GTAAAGGCAGCAGATCTAAATAATA
- Phenotype
-
MIM: 615289
MIM: 608134
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MISP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139343799] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MISP
- Gene Name
- mitotic spindle positioning
- Gene
- PALM
- Gene Name
- paralemmin
There are no transcripts associated with this gene.
View Full Product Details