Product Details

SNP ID
rs7268783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45111982 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTCTGTGACTTTACCTCTGTGTC[C/T]GCCCCTCCCCCTCAGCCCCCTGCCT
Phenotype
MIM: 605161
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WFDC5 PubMed Links

Gene Details

Gene
WFDC5
Gene Name
WAP four-disulfide core domain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145652.3 Intron NP_663627.1
XM_011528601.1 Intron XP_011526903.1
XM_011528602.1 Intron XP_011526904.1

View Full Product Details