Product Details

SNP ID
rs7351730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37256068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAACTAAGAATAATAGTAGCTGAC[A/G]CTACCAAGGGCATCTGTGTGCCCAG
Phenotype
MIM: 139190
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GHRH PubMed Links

Gene Details

Gene
GHRH
Gene Name
growth hormone releasing hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184731.2 Intron NP_001171660.1
NM_021081.5 Intron NP_066567.1
XM_011528784.2 Intron XP_011527086.1
XM_011528788.2 Intron XP_011527090.1

View Full Product Details