Product Details

SNP ID
rs11557000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1118804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCTGGAGGTACTGTTCGCATCG[C/G]CAGCGCCGGCCATCACCTGCAGGCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC105372493 PubMed Links

Gene Details

Gene
LOC105372493
Gene Name
uncharacterized LOC105372493
There are no transcripts associated with this gene.

Gene
PSMF1
Gene Name
proteasome inhibitor subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323407.1 212 Intron NP_001310336.1
NM_001323408.1 212 Missense Mutation CCA,GCA P11A NP_001310337.1
NM_001323409.1 212 Missense Mutation CCA,GCA P11A NP_001310338.1
NM_001323410.1 212 Missense Mutation CCA,GCA P11A NP_001310339.1
NM_006814.4 212 Missense Mutation CCA,GCA P11A NP_006805.2
NM_178578.3 212 Missense Mutation CCA,GCA P11A NP_848693.2

View Full Product Details