Product Details

SNP ID
rs11553684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:37065618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAAAGTTCTTTGGCTGCAAGAAA[A/G]AACATTTGGTTTACTTCACTAATAG
Phenotype
MIM: 605938 MIM: 602259
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105372795 PubMed Links

Gene Details

Gene
LOC105372795
Gene Name
uncharacterized LOC105372795
There are no transcripts associated with this gene.

Gene
PIGP
Gene Name
phosphatidylinositol glycan anchor biosynthesis class P
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320480.1 550 Silent Mutation TTC,TTT F123F NP_001307409.1
NM_016430.3 550 Silent Mutation TTC,TTT F97F NP_057514.2
NM_153681.2 550 Silent Mutation TTC,TTT F147F NP_710148.1
NM_153682.2 550 Silent Mutation TTC,TTT F123F NP_710149.1
XM_005260990.4 550 Intron XP_005261047.1
XM_011529596.2 550 Silent Mutation TTC,TTT F123F XP_011527898.1
XM_017028364.1 550 Silent Mutation TTC,TTT F123F XP_016883853.1
XM_017028365.1 550 Silent Mutation TTC,TTT F97F XP_016883854.1
Gene
TTC3
Gene Name
tetratricopeptide repeat domain 3
There are no transcripts associated with this gene.

View Full Product Details