Product Details

SNP ID
rs11544946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:102153944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTCAAGATTCAAGGCATAAGGC[A/C]AAACTCATCATGATGATCATGGTGA
Phenotype
MIM: 300693
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BEX5 PubMed Links

Gene Details

Gene
BEX5
Gene Name
brain expressed X-linked 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012978.2 624 Missense Mutation GGC,TGC G108C NP_001012996.1
NM_001159560.1 624 Missense Mutation GGC,TGC G108C NP_001153032.1

View Full Product Details