Product Details

SNP ID
hCV44036171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:81118527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAGAAAAGAAAAGGAAAAAAAT[A/C]ATATTTGAAGAAAGAACGCTAAAAA
Phenotype
MIM: 300385
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HMGN5 PubMed Links

Gene Details

Gene
HMGN5
Gene Name
high mobility group nucleosome binding domain 5
There are no transcripts associated with this gene.

View Full Product Details