Product Details

SNP ID
hCV44045859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:68661849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGAACCTTACCACTAACTCTGG[G/T]CTCCCTTTGTTTTTCCTTCTTTCTT
Phenotype
MIM: 300689
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
STARD8 PubMed Links

Gene Details

Gene
STARD8
Gene Name
StAR related lipid transfer domain containing 8
There are no transcripts associated with this gene.

View Full Product Details