Product Details

SNP ID
rs5975629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135902382 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCTAATAGGCATGTAAAATTCAA[C/T]TGACTCTACATGCTCAAATTTCTGA
Phenotype
MIM: 300359
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CT45A10 PubMed Links

Gene Details

Gene
CT45A10
Gene Name
cancer/testis antigen family 45, member A10
There are no transcripts associated with this gene.

Gene
SAGE1
Gene Name
sarcoma antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018666.2 Intron NP_061136.2
XM_017029621.1 Intron XP_016885110.1
XM_017029622.1 Intron XP_016885111.1
XM_017029623.1 Intron XP_016885112.1
XM_017029624.1 Intron XP_016885113.1
XM_017029625.1 Intron XP_016885114.1
XM_017029626.1 Intron XP_016885115.1
XM_017029627.1 Intron XP_016885116.1

View Full Product Details