Product Details

SNP ID
rs5030581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109692709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTCATCTAATCCCATGGCTTCCA[C/T]GACCACCTCCTGATTTATTTCTCCA
Phenotype
MIM: 601532
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASP6 PubMed Links

Gene Details

Gene
CASP6
Gene Name
caspase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001226.3 Intron NP_001217.2
NM_032992.2 Intron NP_116787.1
Gene
MCUB
Gene Name
mitochondrial calcium uniporter dominant negative beta subunit
There are no transcripts associated with this gene.

View Full Product Details