Product Details

SNP ID
rs12760265
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:8321783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATAGACAGATGAATAGATGAATG[C/G]AAGAGAGGATGTGTGGAAGGATGGA
Phenotype
MIM: 605763
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC45A1 PubMed Links

Gene Details

Gene
SLC45A1
Gene Name
solute carrier family 45 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080397.2 Intron NP_001073866.2
XM_011541530.1 Intron XP_011539832.1
XM_011541531.1 Intron XP_011539833.1

View Full Product Details