Product Details

SNP ID
rs10177573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112286363 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCTGGGTTTCCAAAAATATGTCCT[A/G]TTCGGCTGGGTTGAGGGCATAGTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZC3H6 PubMed Links

Gene Details

Gene
ZC3H6
Gene Name
zinc finger CCCH-type containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198581.2 Intron NP_940983.2
XM_006712519.2 Intron XP_006712582.1

View Full Product Details