Product Details

SNP ID
rs13032765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68473282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGTTGGAGTCATACAGTACATA[A/C]CCTTTTCAGGTTTCACTTAGTAATA
Phenotype
MIM: 611035
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APLF PubMed Links

Gene Details

Gene
APLF
Gene Name
aprataxin and PNKP like factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173545.2 Intron NP_775816.1
Gene
FBXO48
Gene Name
F-box protein 48
There are no transcripts associated with this gene.

View Full Product Details